Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Boc-amino)piperidine ≥97%

Supplier: Matrix Scientific
Synonyms: tert-butyl N-(piperidin-4-yl)carbamatetert-Butyl piperidin-4-ylcarbamate4-N-BOC-Aminopiperidine4-N-(tert-Butoxycarbonyl)aminopiperidine

035820-25G
101840-134EA 67.69 USD InStock
101840-134
4-(Boc-amino)piperidine ≥97%
4-(Boc-amino)piperidine
Hazard: Irritant


Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 155…160 °C
MDL Number: MFCD00798171
CAS Number: 73874-95-0
Order Now
Specification Test Results
Melting point 155-160°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty