Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-amino-3-nitrobenzoate ≥95%

Supplier: Matrix Scientific
Synonyms: Methyl 4-amino-3-nitrobenzoate

042844-1G
101857-826EA 15.38 USD InStock
101857-826
Methyl-4-amino-3-nitrobenzoate ≥95%
Methyl 4-amino-3-nitrobenzoate
Hazard: Irritant


Formula: C₈H₈N₂O₄
MW: 196.16 g/mol
Melting Pt: 204…206 °C
MDL Number: MFCD00017562
CAS Number: 3987-92-6
Order Now
Specification Test Results
Melting point 204-206°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty