Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-6-iodopyridazine ≥95%

Supplier: Matrix Scientific

044236-1G
101864-870EA 149.23 USD InStock
101864-870
3-Chloro-6-iodopyridazine ≥95%
3-Chloro-6-iodopyridazine
Hazard: Irritant


Formula: C₄H₂ClIN₂
MW: 240.43 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD08275187
CAS Number: 135034-10-5
Order Now
Specification Test Results
Melting point 118-120°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty