Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Docosanal ≥98%

Supplier: Matrix Scientific

064652-1G
101928-104EA 384.62 USD InStock
101928-104
Docosanal ≥98%
Docosanal
Hazard: Irritant/Refrigerate


Formula: C₂₂H₄₄O
MW: 324.593 g/mol
Melting Pt: 56…57 °C
MDL Number: MFCD07780595
CAS Number: 57402-36-5
Order Now
Specification Test Results
Melting point 56-57°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty