Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-Di-p-toluoyl-D-tartaric acid ≥98%

Supplier: Matrix Scientific
Synonyms: (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric AcidDi-p-toluoyl-D-tartaric AcidDi-p-toluoyl-D-tartaric acid

067065-100G
101933-114EA 69.23 USD InStock
101933-114
(+)-Di-p-toluoyl-D-tartaric acid ≥98%
(+)-Di-p-toluoyl-D-tartaric acid
TCA listed: T
Hazard: Irritant


Formula: C₂₀H₁₈O₈
MW: 386.36 g/mol
Melting Pt: 169…171 °C
MDL Number: MFCD00008552
CAS Number: 32634-68-7
Order Now
Specification Test Results
Melting point 169-171°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty