Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Bromoheptanoic acid ≥97%

Supplier: Matrix Scientific
Synonyms: 7-Bromoenanthic Acid

007518-25G 007518-5G
101792-972EA 238.46 USD InStock
101792-972 101792-970
7-Bromoheptanoic acid ≥97%
7-Bromoheptanoic acid
Hazard: Irritant/Refrigerate


Formula: C₇H₁₃BrO₂
MW: 209.08 g/mol
Boiling Pt: 145 °C (03 mmHg)
Melting Pt: 32 °C
MDL Number: MFCD00037800
CAS Number: 30515-28-7
Order Now
Specification Test Results
Boiling point 145°C/03mm
Melting point 32°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty