Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2,4,6-trifluoropyrimidine ≥98%

Supplier: Matrix Scientific

008726-100G
101795-302EA 383.08 USD InStock
101795-302
5-Chloro-2,4,6-trifluoropyrimidine ≥98%
5-Chloro-2,4,6-trifluoropyrimidine
Hazard: Irritant/Moisture Sensitive/Lachrymator


Formula: C₄ClF₃N₂
Boiling Pt: 116 °C
Density: 1.626
MDL Number: MFCD00191933
CAS Number: 697-83-6
Order Now
Specification Test Results
Boiling point 116°C
Density 1.626
ND20 1.439

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty