Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-3-(2-bromoethoxy)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

047283-1G
101879-296EA 350.77 USD InStock
101879-296
Methyl-3-(2-bromoethoxy)benzenecarboxylate ≥95%
Methyl-3-(2-bromoethoxy)benzenecarboxylate
Hazard: Irritant


Formula: C₁₀H₁₁BrO₃
MW: 259.11 g/mol
Boiling Pt: 110 °C (0.05 mmHg)
MDL Number: MFCD06204041
CAS Number: 59516-96-0
Order Now
Specification Test Results
Boiling point 110°C/0.05m

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty