Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethyl)cyclohexanone ≥97%

Supplier: Matrix Scientific

005489-1G 005489-5G
101789-044EA 304.62 USD InStock
101789-044 101789-042
4-(Trifluoromethyl)cyclohexanone ≥97%
4-(Trifluoromethyl)cyclohexanone
Hazard: Irritant


Formula: C₇H₉F₃O
MW: 166.14
Boiling Pt: 67…68 °C (16 mmHg)
MDL Number: MFCD00102145
UN: 1224
ADR: 3,III
Order Now
Specification Test Results
Boiling point 67-68°C/16m

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty