Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Trifluoromethyl)cyclohexanone ≥97%

Supplier: Matrix Scientific

005497-5G 005497-1G
101789-058EA 169.23 USD InStock
101789-058 101789-060
2-(Trifluoromethyl)cyclohexanone ≥97%
2-(Trifluoromethyl)cyclohexanone
Hazard: Flammable


Formula: C₇H₉F₃O
MW: 166.14
Boiling Pt: 71…73 °C (12 mmHg)
MDL Number: MFCD00102125
UN: 1224
ADR: 3,III
Order Now
Specification Test Results
Boiling point 74°C/18mm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty