Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-(Boc-amino)piperidine ≥95%

Supplier: Matrix Scientific
Synonyms: 3-Aminopiperidine, 3-BOC protectedtert-Butyl piperidin-3-ylcarbamate

036768-5G
101842-042EA 58.46 USD InStock
101842-042
(±)-3-(Boc-amino)piperidine ≥95%
(±)-3-(Boc-amino)piperidine
Hazard: Irritant


Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD03839941
CAS Number: 172603-05-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty