Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-N-methylacetamide ≥95%

Supplier: Matrix Scientific

040456-25G 040456-5G
101849-550EA 136.92 USD InStock
101849-550 101849-548
2-Chloro-N-methylacetamide ≥95%
2-Chloro-N-methylacetamide
Hazard: Irritant


Formula: C₃H₆ClNO
MW: 107.54 g/mol
MDL Number: MFCD00018913
CAS Number: 96-30-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty