Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Difluoromethyl)aniline ≥97%

Supplier: Matrix Scientific

058600-1G
101915-080EA 167.69 USD InStock
101915-080
3-(Difluoromethyl)aniline ≥97%
3-(Difluoromethyl)aniline
Hazard: Irritant-Harmful/Refrigerate


Formula: C₇H₇F₂N
MW: 143.137 g/mol
MDL Number: MFCD18384830
CAS Number: 368-99-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty