Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(4-Bromophenyl)-2-pyrrolidinone

Supplier: Matrix Scientific
Synonyms: 1-(4-Bromophenyl)pyrrolidin-2-one

063443-500MG
101925-726EA 484.62 USD InStock
101925-726
1-(4-Bromophenyl)-2-pyrrolidinone
1-(4-Bromophenyl)-2-pyrrolidinone
Hazard: Irritant


Formula: C₁₀H₁₀BrNO
MW: 240.10 g/mol
MDL Number: MFCD00138502
CAS Number: 7661-32-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty