Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-4-methoxybenzyl bromide ≥98%

Supplier: Matrix Scientific
Synonyms: 4-(Bromomethyl)-2-fluoro-1-methoxybenzene

008958-25G 008958-5G
101795-932EA 84.62 USD InStock
101795-932 101795-934
3-Fluoro-4-methoxybenzyl bromide ≥98%
3-Fluoro-4-methoxybenzyl bromide
Hazard: Irritant/Refrigerate/Lachrymator


Formula: C₈H₈BrFO
MW: 219.05 g/mol
Boiling Pt: 149…155 °C (25…28 Torr)
Melting Pt: 42 °C
MDL Number: MFCD00671768
CAS Number: 331-61-3
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty