Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethynyl-benzoic acid sodium salt

Supplier: Matrix Scientific
Synonyms: Sodium 4-ethynylbenzoate

010261-500MG
101798-184EA 92.31 USD InStock
101798-184
4-Ethynyl-benzoic acid sodium salt
4-Ethynyl-benzoic acid sodium salt
Hazard: Irritant


Formula: C₉H₅NaO₂
MW: 168.13 g/mol
MDL Number: MFCD03426026
CAS Number: 144693-65-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty