Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butylthiazol-2-ylamine

Supplier: Matrix Scientific
Synonyms: 2-Amino-4-(tert-butyl)-1,3-thiazole

010725-500MG
101799-086EA 267.69 USD InStock
101799-086
4-tert-Butylthiazol-2-ylamine
4-tert-Butylthiazol-2-ylamine
Hazard: Irritant


Formula: C₇H₁₂N₂S
MW: 156.25 g/mol
Boiling Pt: 141…143 °C (20 Torr)
Melting Pt: 98…99 °C
MDL Number: MFCD00051644
CAS Number: 74370-93-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty