Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-N,N-dimethylbenzamide

Supplier: Matrix Scientific

026042-500MG
101821-488EA 66.15 USD InStock
101821-488
4-Amino-N,N-dimethylbenzamide
4-Amino-N,N-dimethylbenzamide
Hazard: Irritant


Formula: C₉H₁₂N₂O
MW: 164.21 g/mol
MDL Number: MFCD00523648
CAS Number: 6331-71-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty