Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-2-amino-5-phenylthiophene-3-carboxylate

Supplier: Matrix Scientific
Synonyms: Ethyl 2-amino-5-phenylthiophene-3-carboxylate

029201-1G
101827-676EA 89.23 USD InStock
101827-676
Ethyl-2-amino-5-phenylthiophene-3-carboxylate
Ethyl-2-amino-5-phenylthiophene-3-carboxylate
CAS: 4815-34-3
MDL Number: MFCD01829801
Molecular Formula: C13H13NO2S
Molecular Weight: 247,32
Hazard: Irritant


Formula: C₁₃H₁₃NO₂S
MW: 247.32 g/mol
MDL Number: MFCD01829801
CAS Number: 4815-34-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty