Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine

Supplier: Matrix Scientific
Synonyms: N-Fmoc-pentafluoro-L-phenylalanineFmoc-Phe(F5)-OH

004698-1G
101787-690EA 264.62 USD InStock
101787-690
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine
CAS: 205526-32-5
MDL Number: MFCD00270612
Molecular Formula: C24H16F5NO4
Molecular Weight: 477,39
Hazard: Irritant/Refrigerate


Formula: C₂₄H₁₆F₅NO₄
MW: 477.39 g/mol
MDL Number: MFCD00270612
CAS Number: 205526-32-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty