Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Trifluoro-2,4,6-triiodobenzene ≥98%

Supplier: Matrix Scientific

011263-1G 011263-5G
101800-268EA 492.31 USD InStock
101800-268 101800-266
1,3,5-Trifluoro-2,4,6-triiodobenzene ≥98%
1,3,5-Trifluoro-2,4,6-triiodobenzene
Hazard: Irritant


Formula: C₆F₃I₃
MW: 509.77 g/mol
Melting Pt: 152 °C
MDL Number: MFCD06248899
CAS Number: 84322-56-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty