Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Octahydro-2H-pyrido[1,2-a]pyrazine

Supplier: Matrix Scientific
Synonyms: 1,4-Diazabicyclo[4.4.0]decane

011636-1G
101801-030EA 643.08 USD InStock
101801-030
Octahydro-2H-pyrido[1,2-a]pyrazine
Octahydro-2H-pyrido[1,2-a]pyrazine
CAS: 4430-75-5
MDL Number: MFCD01098257
Molecular Formula: C8H16N2
Molecular Weight: 140,23
Boiling Point: 84°C/12 Torr
Hazard: Irritant


Formula: C₈H₁₆N₂
MW: 140.23 g/mol
Boiling Pt: 84 °C (12 Torr)
MDL Number: MFCD01098257
CAS Number: 4430-75-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty