Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Methoxy[1,1'-biphenyl]-3-amine

Supplier: Matrix Scientific
Synonyms: 3-Amino-4'-methoxybiphenyl hydrochloride

021703-500MG
101812-582EA 69.23 USD InStock
101812-582
4'-Methoxy[1,1'-biphenyl]-3-amine
4'-Methoxy[1,1'-biphenyl]-3-amine
Hazard: Irritant


Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
MDL Number: MFCD03424668
CAS Number: 53059-28-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty