Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Isocyanato-1-methoxy-4-methylbenzene

Supplier: Matrix Scientific
Synonyms: 2-Methoxy-5-methylphenyl isocyanate

030176-500MG
101829-634EA 146.15 USD InStock
101829-634
2-Isocyanato-1-methoxy-4-methylbenzene
2-Isocyanato-1-methoxy-4-methylbenzene
Hazard: Irritant


Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00013869
CAS Number: 59741-04-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty