Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate

Supplier: Matrix Scientific
Synonyms: N-Fmoc-L-aspartic Acid 4-Benzyl EsterFmoc-Asp(OBzl)-OHN-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-Benzyl Ester4-Benzyl N-Fmoc-L-aspartate

041298-5G 041298-25G
101851-260EA 304.62 USD InStock
101851-260 101851-258
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate
Hazard: Irritant


Formula: C₂₆H₂₃NO₆
MW: 445.47 g/mol
MDL Number: MFCD00065630
CAS Number: 86060-84-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty