Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fmoc-d-asp-otbu

Supplier: Matrix Scientific
Synonyms: Fmoc-D-Asp-OtBuN-Fmoc-D-aspartic acid 1-tert-butyl ester

041304-1G 041304-5G
101851-284EA 1176.92 USD InStock
101851-284 101851-282
Fmoc-d-asp-otbu
Fmoc-d-asp-otbu
Hazard: Irritant


Formula: C₂₃H₂₅NO₆
MW: 411.454 g/mol
MDL Number: MFCD00237011
CAS Number: 134098-70-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty