Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-(Fmoc-amino)valeric acid ≥95%

Supplier: Matrix Scientific
Synonyms: Fmoc-5-Ava-OH

041394-5G
101851-604EA 152.31 USD InStock
101851-604
5-(Fmoc-amino)valeric acid ≥95%
5-(Fmoc-amino)valeric acid
Hazard: Irritant


Formula: C₂₀H₂₁NO₄
MW: 339.391 g/mol
MDL Number: MFCD00235889
CAS Number: 123622-48-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty