Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-D-ABU-OH

Supplier: Matrix Scientific
Synonyms: (R)-2-(Boc-amino)butyric acid

041454-1G
101851-842EA 15.38 USD InStock
101851-842
Boc-D-ABU-OH
Boc-D-ABU-OH
CAS: 45121-22-0
MDL Number: MFCD00270335
Molecular Formula: C9H17NO4
Molecular Weight: 203,24
Hazard: Irritant


Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
MDL Number: MFCD00270335
CAS Number: 45121-22-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty