Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-D-4-Cyanophenylalanine

Supplier: Matrix Scientific
Synonyms: Boc-D-Phe(4-CN)-OHBoc-4-cyano-D-Phe-OHN-Boc-4-cyano-D-phenylalanine

041885-1G 041885-5G
101853-592EA 1680 USD InStock
101853-592 101853-590
Boc-D-4-Cyanophenylalanine
Boc-D-4-Cyanophenylalanine
CAS: 146727-62-0
MDL Number: MFCD00672527
Molecular Formula: C15H18N2O4
Molecular Weight: 290,32
Hazard: Irritant


Formula: C₁₅H₁₈N₂O₄
MW: 290.319 g/mol
MDL Number: MFCD00672527
CAS Number: 146727-62-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty