Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific
Synonyms: (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acidBoc-Cis-4-F-Pro-OH(2S,4S)-1-[(tert-butoxy)carbonyl]-4-fluoropyrrolidine-2-carboxylic acidN-(tert-Butoxycarbonyl)-cis-4-fluoro-L-proline(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acidN-Boc-cis-4-fluoro-L-proline(2S,4S)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid

042033-1G
101854-182EA 95.38 USD InStock
101854-182
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
Hazard: Irritant


Formula: C₁₀H₁₆FNO₄
MW: 233.24 g/mol
MDL Number: MFCD04973957
CAS Number: 203866-13-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty