Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-1,3-propanediamine ≥95%

Supplier: Matrix Scientific
Synonyms: N-Boc-1,3-propanediaminetert-Butyl (3-aminopropyl)carbamatetert-Butyl N-(3-Aminopropyl)carbamateN-Boc-1,3-diaminopropaneN-(3-Aminopropyl)carbamic Acid tert-Butyl EsterN-1-Boc-1,3-diaminopropane·HClN-(tert-Butoxycarbonyl)-1,3-propanediamine

046721-5G
101876-732EA 29.23 USD InStock
101876-732
N-Boc-1,3-propanediamine ≥95%
N-Boc-1,3-propanediamine
Hazard: Corrosive/Air Sensitive/Refrigerate


Formula: C₈H₁₈N₂O₂
MW: 174.244 g/mol
Melting Pt: ∼ 22 °C
MDL Number: MFCD00210021
CAS Number: 75178-96-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty