Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromomethyl-2,2-difluorocyclopropane ≥97%

Supplier: Matrix Scientific

059629-1G
101918-672EA 400 USD InStock
101918-672
1-Bromomethyl-2,2-difluorocyclopropane ≥97%
1-Bromomethyl-2,2-difluorocyclopropane
Hazard: Flammable/Refrigerate


Formula: C₄H₅BrF₂
MW: 170.985 g/mol
Boiling Pt: 108…109 °C
MDL Number: MFCD06797749
CAS Number: 77613-65-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty