Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Boc-3-methylamino azetidine hydrochloride ≥95%

Supplier: Matrix Scientific

059918-5G 059918-1G
101919-450EA 335.38 USD InStock
101919-450 101919-448
3-Boc-3-methylamino azetidine hydrochloride ≥95%
3-Boc-3-methylamino azetidine hydrochloride
Hazard: Irritant


Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD12963843
CAS Number: 577777-20-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty