Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-[1-(3-Aminophenyl)ethyl]acetamide compound with sulfuric acid (2:1)

Supplier: Matrix Scientific

060197-500MG
101919-598EA 364.62 USD InStock
101919-598
N-[1-(3-Aminophenyl)ethyl]acetamide compound with sulfuric acid (2:1)
N-[1-(3-Aminophenyl)ethyl]acetamide compound with sulfuric acid
Hazard: Irritant


Formula: C₂₀H₃₀N₄O₆S
MDL Number: MFCD18252914
CAS Number: 103394-66-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty