Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(1,3-Benzodioxol-5-ylmethyl)guanidine compound with sulfuric acid (2:1)

Supplier: Matrix Scientific

060423-500MG
101920-032EA 243.08 USD InStock
101920-032
N-(1,3-Benzodioxol-5-ylmethyl)guanidine compound with sulfuric acid (2:1)
N-(1,3-Benzodioxol-5-ylmethyl)guanidine compound with sulfuric acid
Hazard: Irritant


Formula: C₁₈H₂₄N₆O₈S
MDL Number: MFCD01746761
CAS Number: 17228-46-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty