Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-1,3-dimethyluracil

Supplier: Matrix Scientific
Synonyms: 6-Chloro-1,3-dimethylpyrimidine-2,4(1H,3H)-dione

065836-5G
101930-452EA 23.08 USD InStock
101930-452
6-Chloro-1,3-dimethyluracil
6-Chloro-1,3-dimethyluracil
Hazard: Irritant


Formula: C₆H₇ClN₂O₂
MW: 174.59 g/mol
Melting Pt: 109…111 °C
MDL Number: MFCD00038066
CAS Number: 6972-27-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty