Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethynyltributyltin ≥95%

Supplier: Matrix Scientific
Synonyms: Ethynyltri-n-butylstannaneTributylethynylstannane

069498-1G 069498-5G
101940-464EA 56.92 USD InStock
101940-464 101940-466
Ethynyltributyltin ≥95%
Ethynyltributyltin
Hazard: Toxic/Moisture Sensitive


Formula: C₁₄H₂₈Sn
MW: 315.09
Boiling Pt: 70 °C (0.2 mmHg)
Density: 1.08
Flash Pt: 73 °C
MDL Number: MFCD00009420
CAS Number: 994-89-8
UN: 2788
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty