Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(4-Fluorophenyl)piperidin-4-ol

Supplier: Matrix Scientific
Synonyms: 4-(4-Fluorophenyl)-4-hydroxypiperidine

012457-500MG
101802-612EA 273.85 USD InStock
101802-612
4-(4-Fluorophenyl)piperidin-4-ol
4-(4-Fluorophenyl)piperidin-4-ol
Hazard: Irritant


Formula: C₁₁H₁₄FNO
Melting Pt: 116…117 °C
MDL Number: MFCD00023146
CAS Number: 3888-65-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty