Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethylsulfony)phenylacetic acid ≥98%

Supplier: Matrix Scientific

032361-1G
101833-568EA 120 USD InStock
101833-568
4-(Trifluoromethylsulfony)phenylacetic acid ≥98%
4-(Trifluoromethylsulfony)phenylacetic acid
Hazard: Irritant


Formula: C₉H₇F₃O₄S
MW: 268.21 g/mol
Melting Pt: 126…127 °C
MDL Number: MFCD04973012
CAS Number: 1099597-82-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty