Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

035666-5G 035666-1G
101839-610EA 21.54 USD InStock
101839-610 101839-612
3-Amino-1H-pyrazole-4-carboxylic acid ≥95%
3-Amino-1H-pyrazole-4-carboxylic acid
Hazard: Irritant/Moisture Sensitive/Refrigerate


Formula: C₄H₅N₃O₂
MW: 127.10 g/mol
Melting Pt: 135 °C. (dec.)
MDL Number: MFCD00005239
CAS Number: 41680-34-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty