Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Phthaloylglycine

Supplier: Matrix Scientific
Synonyms: (1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetic acid

035821-100G
101840-136EA 72.31 USD InStock
101840-136
N-Phthaloylglycine
N-Phthaloylglycine
Hazard: Irritant


Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
MDL Number: MFCD00005900
CAS Number: 4702-13-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty