Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-m-Coumaric acid

Supplier: Matrix Scientific
Synonyms: (2E)-3-(3-Hydroxyphenyl)acrylic acidm-Coumaric acid(E)-3-(3-Hydroxyphenyl)acrylic acidm-Hydroxycinnamic acid, predominantly transtrans-3-Hydroxycinnamic acid

037614-25G
101843-800EA 41.54 USD InStock
101843-800
trans-m-Coumaric acid
trans-m-Coumaric acid
CAS: 14755-02-3
MDL Number: MFCD00004390
Molecular Formula: C9H8O3
Molecular Weight: 164,16
Hazard: Irritant


Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 192…196 °C
MDL Number: MFCD00004390
CAS Number: 14755-02-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty