Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hydroxyindol-3-ylacetic acid

Supplier: Matrix Scientific
Synonyms: 2-(5-Hydroxy-1H-indol-3-yl)acetic acid5-Hydroxyindole-3-acetic acid

037615-500MG
101843-804EA 290.77 USD InStock
101843-804
5-Hydroxyindol-3-ylacetic acid
5-Hydroxyindol-3-ylacetic acid
Hazard: Irritant


Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
MDL Number: MFCD00005639
CAS Number: 54-16-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty