Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Boc-aminopyrrolidine ≥95%

Supplier: Matrix Scientific
Synonyms: 3-Boc-Aminopyrrolidine1,1-Dimethylethyl3-pyrrolidinylcarbamate3-(BOC-amino)pyrrolidine2-Methyl-2-propanyl 3-pyrrolidinylcarbamate

037634-1G
101843-848EA 16.92 USD InStock
101843-848
3-Boc-aminopyrrolidine ≥95%
3-Boc-aminopyrrolidine
Hazard: Irritant


Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
MDL Number: MFCD00059040
CAS Number: 99724-19-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty