Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fmoc-L-β-homoglutamic acid-d-t-butyl ester

Supplier: Matrix Scientific
Synonyms: Fmoc-β-HoGlu(OtBu)-OHFmoc-β-HomoGlu(OtBu)-OH

042366-1G 042366-250MG
101855-522EA 773.85 USD InStock
101855-522 101855-520
Fmoc-L-β-homoglutamic acid-d-t-butyl ester
Fmoc-L-beta-homoglutamic acid-d-t-butyl ester
Hazard: Irritant


Formula: C₂₅H₂₉NO₆
MW: 439.51 g/mol
MDL Number: MFCD01863053
CAS Number: 203854-49-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty