Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluoxetine hydrochloride ≥95%

Supplier: Matrix Scientific
Synonyms: N-Methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propylamine Hydrochloride

047891-5G
101881-876EA 223.08 USD InStock
101881-876
Fluoxetine hydrochloride ≥95%
Fluoxetine hydrochloride
Purity: 95+%
CAS: 56296-78-7
MDL Number: MFCD00214288
Molecular Formula: C17H19ClF3NO
Molecular Weight: 345,8
Hazard: Irritant


Formula: C₁₇H₁₈F₃NO·HCl
MW: 345.79 g/mol
Melting Pt: 158…159 °C
MDL Number: MFCD00214288
CAS Number: 56296-78-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty