Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Formyl-L-leucine ≥90%

Supplier: Matrix Scientific
Synonyms: N-Formyl-Leu-OH

048721-1G
101885-012EA 53.85 USD InStock
101885-012
N-Formyl-L-leucine ≥90%
N-Formyl-L-leucine
Purity: 90%
CAS: 6113-61-7
MDL Number: MFCD00055861
Molecular Formula: C7H13NO3
Molecular Weight: 159,19
Hazard: Irritant


Formula: C₇H₁₃NO₃
MW: 159.19 g/mol
MDL Number: MFCD00055861
CAS Number: 6113-61-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty