Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride ≥97%

Supplier: Matrix Scientific
Synonyms: Ethyl 2-amino-3,3,3-trifluoropropionate hydrochlorideEthyl 2-amino-3,3,3-trifluoropropanoate hydrochloride

055423-1G
101909-230EA 203.08 USD InStock
101909-230
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride ≥97%
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride
Hazard: Irritant


Formula: C₅H₉ClF₃NO₂
MW: 207.58 g/mol
MDL Number: MFCD14525479
CAS Number: 193140-71-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty