Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sudan black B

Supplier: Matrix Scientific
Synonyms: Ceres black BNSolvent Black 3CI 26150Fat Black HB

058387-500MG
101914-706EA 306.15 USD InStock
101914-706
Sudan black B
Sudan black B
Hazard: Irritant


Formula: C₂₉H₂₄N₆
MW: 456.55 g/mol
MDL Number: MFCD00006919
CAS Number: 4197-25-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty