Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Methoxycoumarin-4-acetic acid

Supplier: Matrix Scientific
Synonyms: 2-(7-Methoxy-2-oxo-2H-chromen-4-yl)acetic acid(7-Methoxycoumarin-4-yl)acetic acidMca7-Methoxy-2-oxo-2H-1-benzopyran-4-acetic acid

064790-500MG
101928-380EA 364.62 USD InStock
101928-380
7-Methoxycoumarin-4-acetic acid
7-Methoxycoumarin-4-acetic acid
Hazard: Irritant


Formula: C₁₂H₁₀O₅
MW: 234.207 g/mol
MDL Number: MFCD00009774
CAS Number: 62935-72-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty